Learn More
TARGETMOL CHEMICALS INC 4-HYDROXYTAMOXIFEN 25MG
SDP
Catalog No. 502579885
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-Hydroxytamoxifen (ICI 79280) is a selective estrogen receptor modulator (SERM). purity: 99%
Catalog No. 50-257-9885 Supplier TARGETMOL CHEMICALS INC Supplier No. T442025MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 4-Hydroxytamoxifen (ICI 79280) is a selective estrogen receptor modulator (SERM). purity: 99%