Learn More
American Radiolabeled Chemicals Inc 4-HYDROXYTAMOXIFEN-3H 250UC
SDP
Catalog No. 5090104809
Encompass_Preferred
4-Hydroxytamoxifen methiodide-N-methyl-3H 250uCi in ethanol, 1 mCi/ml, S.A. 65-87 Ci/mmol, M.W. 529.4, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-04809 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1106250UC
4-Hydroxytamoxifen methiodide-N-methyl-3H 250uCi in ethanol, 1 mCi/ml, S.A. 65-87 Ci/mmol, M.W. 529.4, Shipped in Dry Ice, Exclusive