Learn More
Medchemexpress LLC HY-16950 50mg , 4-Hydroxytamoxifen (Z)-4-Hydroxytamoxifen;trans-4-Hydroxytamoxifen CAS:68047-06-3 Purity:98%
SDP
Catalog No. 501493673
Encompass_Preferred
Medchemexpress, HY-16950 50mg 4-Hydroxytamoxifen (Z)-4-Hydroxytamoxifen;trans-4-Hydroxytamoxifen CAS:68047-06-3 Formula:C26H29NO2 IC50: 3.3 nM (Oestrogen receptor) CRISPR/Cas9 Purity:98% 4-Hydroxytamoxifen, an active metabolite of tamoxifen, is a selective estrogen receptor antagonist, with an IC 50 of 3.3 nM for the [ 3 H]oestradiol binding to oestrogen receptor, widely used to treat breast cancer; 4-Hydroxytamoxifen also increases CRISPR/Cas9 -mediated editing frequency. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3673 Supplier Medchemexpress LLC Supplier No. HY1695050MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-16950 50mg 4-Hydroxytamoxifen (Z)-4-Hydroxytamoxifen;trans-4-Hydroxytamoxifen CAS:68047-06-3 Formula:C26H29NO2 IC50: 3.3 nM (Oestrogen receptor) CRISPR/Cas9 Purity:98% 4-Hydroxytamoxifen, an active metabolite of tamoxifen, is a selective estrogen receptor antagonist, with an IC 50 of 3.3 nM for the [ 3 H]oestradiol binding to oestrogen receptor, widely used to treat breast cancer; 4-Hydroxytamoxifen also increases CRISPR/Cas9 -mediated editing frequency. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.