Learn More
Selleck Chemical LLC -4-Hydroxytamoxifen 5mg
SDP
Catalog No. 502173455
Encompass_Preferred
(Z)-4-Hydroxytamoxifen ((Z)-4-OHT, trans-4-Hydroxytamoxifen, 4-Monohydroxytamoxifen) is a selective modulator of estrogen receptor (SERM) with IC50 of 3.3 nM for the [3H]oestradiol binding to oestrogen receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3455 Supplier Selleck Chemical LLC Supplier No. S89565MG
(Z)-4-Hydroxytamoxifen ((Z)-4-OHT, trans-4-Hydroxytamoxifen, 4-Monohydroxytamoxifen) is a selective modulator of estrogen receptor (SERM) with IC50 of 3.3 nM for the [3H]oestradiol binding to oestrogen receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.