Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxytolbutamide | 5719-85-7 | MFCD00144466 | 99.7% | 286.35 g/mol | C12H18N2O4S | 10 MG
SDP

Catalog No. 5000213533
Encompass_Preferred

4-Hydroxytolbutamide is a research-grade metabolite of the antidiabetic tolbutamide, provided as a solid powder for biochemical and pharmacological studies. It is commonly used as an analytical reference standard and in in vitro metabolism experiments involving CYP2C8 and CYP2C9.

  • High purity (99.7%).
  • CAS number 5719-85-7 for unambiguous identification.
  • Molecular weight 286.35 g/mol, suitable for analytical calculations.
  • Solid powder stable when stored at -20°C for long-term storage.
  • Available in small pack sizes for analytical and research use.

Catalog No. 50-002-13533 Supplier Medchemexpress LLC Supplier No. HY10064110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxytolbutamide is a research-grade metabolite of the antidiabetic tolbutamide, provided as a solid powder for biochemical and pharmacological studies. It is commonly used as an analytical reference standard and in in vitro metabolism experiments involving CYP2C8 and CYP2C9.

  • High purity (99.7%).
  • CAS number 5719-85-7 for unambiguous identification.
  • Molecular weight 286.35 g/mol, suitable for analytical calculations.
  • Solid powder stable when stored at -20°C for long-term storage.
  • Available in small pack sizes for analytical and research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.