Learn More
Medchemexpress LLC 4-hydroxytolbutamide | 5719-85-7 | MFCD00144466 | 99.7% | 286.35 g/mol | C12H18N2O4S | 5 MG
SDP
Catalog No. 5000213669
Encompass_Preferred
4-Hydroxytolbutamide is a hydroxylated metabolite of the sulfonylurea tolbutamide, used as a reference compound in studies of drug metabolism and cytochrome P450 enzyme activity. It is employed to investigate biotransformation pathways mediated by CYP2C8 and CYP2C9.
  • Suitable as a reference standard for CYP2C8 and CYP2C9 metabolism studies.
  • High purity (>99%) for analytical and bioanalytical applications.
  • Solid powder form for ease of handling and storage.
  • Stored at -20°C in powder; in solvent store at -80°C for long-term stability.
  • Chemical formula C12H18N2O4S; molecular weight 286.35 g/mol.
Catalog No. 50-002-13669 Supplier Medchemexpress LLC Supplier No. HY1006415MG
4-Hydroxytolbutamide is a hydroxylated metabolite of the sulfonylurea tolbutamide, used as a reference compound in studies of drug metabolism and cytochrome P450 enzyme activity. It is employed to investigate biotransformation pathways mediated by CYP2C8 and CYP2C9.
  • Suitable as a reference standard for CYP2C8 and CYP2C9 metabolism studies.
  • High purity (>99%) for analytical and bioanalytical applications.
  • Solid powder form for ease of handling and storage.
  • Stored at -20°C in powder; in solvent store at -80°C for long-term stability.
  • Chemical formula C12H18N2O4S; molecular weight 286.35 g/mol.