Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Hydroxytryptamine 1mg | 55206-11-6 | 387.41 | C14H21N5O6S | 1 MG
SDP

Catalog No. 5000229362
Encompass_Preferred

4-Hydroxytryptamine creatinine sulfate is the creatinine sulfate salt of 4-hydroxytryptamine (serotonin). It is a tryptamine derivative used as a research reagent and acts as a neurotransmitter agonist; reported activities include inhibition of [3H] norepinephrine uptake and modulation of norepinephrine release in animal studies.

  • High purity: 97.9% (assay).
  • Solid powder form, suitable for analytical and biological studies.
  • Molecular weight 387.41 g·mol⁻¹.
  • Chemical formula C14H21N5O6S.
  • Provided in milligram-scale pack sizes for research use.
  • Documentation available: COA, SDS, and elemental analysis report.

Catalog No. 50-002-29362 Supplier Medchemexpress LLC Supplier No. HY1157621MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxytryptamine creatinine sulfate is the creatinine sulfate salt of 4-hydroxytryptamine (serotonin). It is a tryptamine derivative used as a research reagent and acts as a neurotransmitter agonist; reported activities include inhibition of [3H] norepinephrine uptake and modulation of norepinephrine release in animal studies.

  • High purity: 97.9% (assay).
  • Solid powder form, suitable for analytical and biological studies.
  • Molecular weight 387.41 g·mol⁻¹.
  • Chemical formula C14H21N5O6S.
  • Provided in milligram-scale pack sizes for research use.
  • Documentation available: COA, SDS, and elemental analysis report.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.