Learn More
Medchemexpress LLC 4'-(Imidazol-1-yl) acetophenone | 10041-06-2 | 99.9% | 186.21 | 25 G
SDP
Catalog No. 5000347679
Encompass_Preferred
The product is a selective thromboxane synthetase inhibitor. It is also known by its synonym, (4'-(Imidazol-1-yl) acetophenone).
  • Provided as a solid, white to off-white in appearance.
  • High purity, typically 99.91%.
  • Stable for 3 years at room temperature in continental US.
  • Can be stored in solvent for 2 years at -80°C and 1 year at -20°C.
  • Available in various quantities, from 5 g to 50 g.
Catalog No. 50-003-47679 Supplier Medchemexpress LLC Supplier No. 25GHY109877
The product is a selective thromboxane synthetase inhibitor. It is also known by its synonym, (4'-(Imidazol-1-yl) acetophenone).
  • Provided as a solid, white to off-white in appearance.
  • High purity, typically 99.91%.
  • Stable for 3 years at room temperature in continental US.
  • Can be stored in solvent for 2 years at -80°C and 1 year at -20°C.
  • Available in various quantities, from 5 g to 50 g.