Learn More
Medchemexpress LLC 4-(Imidazol-1-yl)phenol | 10041-02-8 | 160.17 | 5 G
SDP
Catalog No. 5000446319
Encompass_Preferred
4-(Imidazol-1-yl)phenol is a highly potent signal enhancer used in a horseradish peroxidase (HRP)-luminol chemiluminescence (CL) immunoassay.
  • Highly potent signal enhancer
  • Used in HRP-luminol chemiluminescence immunoassays
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-46319 Supplier Medchemexpress LLC Supplier No. HYW0003575G
4-(Imidazol-1-yl)phenol is a highly potent signal enhancer used in a horseradish peroxidase (HRP)-luminol chemiluminescence (CL) immunoassay.
  • Highly potent signal enhancer
  • Used in HRP-luminol chemiluminescence immunoassays
  • Solid appearance
  • White to off-white color