Learn More
Medchemexpress LLC 4-imidazolidinehexanamide, n-(14-amino-3,6,9,12-tetraoxatetradec-1-yl)-5-methyl-2-oxo-, (4R,5S)- | 2306109-91-9 | 99.2% | C20H40N4O6 | 10MG
SDP
Catalog No. 5000259224
Encompass_Preferred
Amine-PEG4-Desthiobiotin is a PEG-based linker bearing a primary amine and a desthiobiotin moiety used to assemble proteolysis-targeting chimeras (PROTACs), affinity probes, and bioconjugates. The reagent supplies a hydrophilic spacer and a reactive handle to link target-binding ligands to recruiting ligands, supporting modular synthesis and downstream affinity-based workflows.
  • Provides a primary amine for straightforward conjugation using standard amide-coupling chemistries.
  • Contains a desthiobiotin moiety for reversible affinity capture with streptavidin-based systems.
  • PEG4 spacer imparts hydrophilicity and flexible linkage between functional groups.
  • High reported purity supports reproducible synthesis and analytical characterization.
  • Stable when stored frozen and in solution under recommended conditions.
Catalog No. 50-002-59224 Supplier Medchemexpress LLC Supplier No. HY13472010MG
Amine-PEG4-Desthiobiotin is a PEG-based linker bearing a primary amine and a desthiobiotin moiety used to assemble proteolysis-targeting chimeras (PROTACs), affinity probes, and bioconjugates. The reagent supplies a hydrophilic spacer and a reactive handle to link target-binding ligands to recruiting ligands, supporting modular synthesis and downstream affinity-based workflows.
  • Provides a primary amine for straightforward conjugation using standard amide-coupling chemistries.
  • Contains a desthiobiotin moiety for reversible affinity capture with streptavidin-based systems.
  • PEG4 spacer imparts hydrophilicity and flexible linkage between functional groups.
  • High reported purity supports reproducible synthesis and analytical characterization.
  • Stable when stored frozen and in solution under recommended conditions.