Learn More
Medchemexpress LLC 4-Imidazolidinehexanamide, N-[2-[(2-iodoacetyl)amino]ethyl]-5-methyl-2-oxo-, (4R,5S)- | 2924824-04-2 | MFCD34641797 | 99.5% | 424.28 | C14H25IN4O3 | 10 MG
SDP
Catalog No. 5000287218
Encompass_Preferred
Desthiobiotin-iodoacetamide is an iodoacetamide-derived desthiobiotin reagent used in bioconjugation and chemoproteomics. It selectively reacts with cysteine residues to label and enrich proteins, and can be employed in antibody-drug conjugate (ADC) linker chemistry or as a probe in protein labeling workflows. The compound is supplied as a solid powder and is intended for research use only.
  • Reacts with cysteine residues for selective labeling.
  • Enables ADC linker formation for conjugation workflows.
  • Facilitates affinity enrichment and proteomic profiling.
  • High reported purity for consistent experimental performance.
  • Stable solid form with recommended cold storage for longevity.
Catalog No. 50-002-87218 Supplier Medchemexpress LLC Supplier No. HY15023010MG
Desthiobiotin-iodoacetamide is an iodoacetamide-derived desthiobiotin reagent used in bioconjugation and chemoproteomics. It selectively reacts with cysteine residues to label and enrich proteins, and can be employed in antibody-drug conjugate (ADC) linker chemistry or as a probe in protein labeling workflows. The compound is supplied as a solid powder and is intended for research use only.
  • Reacts with cysteine residues for selective labeling.
  • Enables ADC linker formation for conjugation workflows.
  • Facilitates affinity enrichment and proteomic profiling.
  • High reported purity for consistent experimental performance.
  • Stable solid form with recommended cold storage for longevity.