Learn More
eMolecules​ 533-48-2 | D-Desthiobiotin | Ambeed | MFCD00213804 | 214.265 | C10H18N2O3 | 97 | C[C@@H]1NC(=O)N[C@@H]1CCCCCC(O)=O | 1g | 603089220
Supplier:  eMolecules​ 603089220
Encompass_Preferred
D-Desthiobiotin | Ambeed | 533-48-2 | MFCD00213804 | 214.265 | C10H18N2O3 | 97.000 | C[C@@H]1NC(=O)N[C@@H]1CCCCCC(O)=O | 1g | 603089220
Catalog No. NC2037700