Learn More
Medchemexpress LLC 4-Imidazolidinone, 2-thioxo- | 503-87-7 | MFCD00005277 | 99.9% | 116.14 | 100 MG
SDP
Catalog No. 5000455057
Encompass_Preferred
2-Thiohydantoin acts as an inhibitor for the corrosion of mild steel in 0.1 M HCl, and its inhibition efficiency is dependent on both concentration and immersion time. It is a solid with a light yellow to brown appearance.
  • Acts as an inhibitor for the corrosion of mild steel in 0.1 M HCl
  • Inhibition efficiency is dependent on concentration
  • Inhibition efficiency is dependent on immersion time
Catalog No. 50-004-55057 Supplier Medchemexpress LLC Supplier No. HYW012896100MG
2-Thiohydantoin acts as an inhibitor for the corrosion of mild steel in 0.1 M HCl, and its inhibition efficiency is dependent on both concentration and immersion time. It is a solid with a light yellow to brown appearance.
  • Acts as an inhibitor for the corrosion of mild steel in 0.1 M HCl
  • Inhibition efficiency is dependent on concentration
  • Inhibition efficiency is dependent on immersion time