Learn More
TRIBIOSCIENCE INC 4-in-1 Aspergillus qPCR Kit, 200 Reactions
SDP
Catalog No. NC2887504
Encompass_Preferred
The 4-in-1 Aspergillus qPCR Kit is designed for identifying 4 Aspergillus species of A. flavus, A. fumigatus, A. niger, and A. terreus in a single PCR reaction using real-time quantitative polymerase chain reaction (qPCR) and probe fluorescence labels.
Catalog No. NC2887504 Supplier TRIBIOSCIENCE INC Supplier No. TBS42025200
The 4-in-1 Aspergillus qPCR Kit is designed for identifying 4 Aspergillus species of A. flavus, A. fumigatus, A. niger, and A. terreus in a single PCR reaction using real-time quantitative polymerase chain reaction (qPCR) and probe fluorescence labels.