Learn More
Opentrons Labworks Inc 4-IN-1 TUBE RACK SET
Catalog No. NC1965773
Encompass
  • Versatile labware accessory for efficient tube organization.
  • Compatible with automated liquid handling systems.
  • Interchangeable tops accommodate various tube sizes.
  • Snap-on design ensures stability during automated processes.
  • Fits directly on OT-2 deck.
  • Components include 2 base stands and 4 tube holder tops.
  • Tube compatibility: 1.5 mL, 2 mL, 15 mL, 50 mL.
  • Not autoclavable.
  • 1 Year warranty.
  • Shipping time: 7 business days.
Catalog No. NC1965773 Supplier Opentrons Labworks Inc Supplier No. 99900030
  • Versatile labware accessory for efficient tube organization.
  • Compatible with automated liquid handling systems.
  • Interchangeable tops accommodate various tube sizes.
  • Snap-on design ensures stability during automated processes.
  • Fits directly on OT-2 deck.
  • Components include 2 base stands and 4 tube holder tops.
  • Tube compatibility: 1.5 mL, 2 mL, 15 mL, 50 mL.
  • Not autoclavable.
  • 1 Year warranty.
  • Shipping time: 7 business days.