Learn More
Ace Glass, Inc. Coupling, makes bead to bead coupling of 4 inch glass process drain pipe, 80in-lbs max torque, max temp400°F (204°C)
SDP
Catalog No. 50976612
Encompass
Catalog No. 50-976-612 Supplier Ace Glass, Inc. Supplier No. 885615