Learn More
Ted Pella Inc Foil Roll, 4", for the Mini Bin Foil Dispenser
SDP
Catalog No. NC1119486
Encompass
Designed exclusively for the Mini Bin Foil Dispenser. Available in two sizes; 4" or 6" width. Both rolls are 50.8 feet (15.48m).
Catalog No. NC1119486 Supplier Ted Pella Inc Supplier No. 8094
Designed exclusively for the Mini Bin Foil Dispenser. Available in two sizes; 4" or 6" width. Both rolls are 50.8 feet (15.48m).