Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Analyzer Corporation Fan, 4 , 4600, 120 V
SDP

Catalog No. NC1982683
Encompass

FAN 4", 120V 4600

Catalog No. NC1982683 Supplier Petroleum Analyzer Corporation Supplier No. 61009115V
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FAN 4", 120V 4600

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.