Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Iodo-1H-imidazole | 71759-89-2 | 193.97 | 10 G
SDP

Catalog No. 5000449039
Encompass_Preferred

4-Iodo-1H-imidazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Appearance: Solid, white to light yellow
  • Purity: 98.47%
  • Molecular weight: 193.97
  • Molecular formula: C3H3IN2
  • Solubility in DMSO: 175 mg/mL

Catalog No. 50-004-49039 Supplier Medchemexpress LLC Supplier No. HYW00280210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Iodo-1H-imidazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Appearance: Solid, white to light yellow
  • Purity: 98.47%
  • Molecular weight: 193.97
  • Molecular formula: C3H3IN2
  • Solubility in DMSO: 175 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.