Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-2-nitroanisole, 98%

Catalog No. AAB2566603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2566603 1 g
AAB2566606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2566603 Supplier Thermo Scientific Chemicals Supplier No. B2566603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 52692-09-8 | C7H6INO3 | 279.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52692-09-8
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.033
MDL Number MFCD08060933
InChI Key WULXGCDMVLQZBT-UHFFFAOYSA-N
Synonym 4-iodo-2-nitroanisole, 4-iodo-1-methoxy-2-nitro-benzene, benzene, 4-iodo-1-methoxy-2-nitro, 2-nitro-4-iodoanisole, acmc-1amtn, 4-methoxy-3-nitroiodobenzene, benzene,4-iodo-1-methoxy-2-nitro, 4-iodanyl-1-methoxy-2-nitro-benzene
PubChem CID 12249866
IUPAC Name 4-iodo-1-methoxy-2-nitrobenzene
SMILES COC1=C(C=C(C=C1)I)[N+](=O)[O-]

Specifications

Melting Point 96°C to 98°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 279.04
Percent Purity 98%
Chemical Name or Material 4-Iodo-2-nitroanisole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.