Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-3,5-dimethyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog No. AAB2064788
Change view
Click to view available options
Quantity:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2064788 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2064788 Supplier Thermo Scientific Chemicals Supplier No. B2064788

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2033-45-6
Odor Odorless
Molecular Formula C5H7IN2
Quantity 2.5g
MDL Number MFCD00040247
Sensitivity Light sensitive
Synonym 3,5-dimethyl-4-iodopyrazole, 4-iodo-3,5-dimethylpyrazole, 3,5-dimethyl-4-iodo-1h-pyrazole, pyrazole, 3,5-dimethyl-4-iodo, 3,5-dimethyl-4-lodopyrazole, 1h-pyrazole, 4-iodo-3,5-dimethyl, zlchem 469, acmc-209f8u, 5-23-05-00159 beilstein handbook reference, ksc490o8h
InChI Key MZZXIXHKDJNBJQ-UHFFFAOYSA-N
SMILES CC1=C(C(=NN1)C)I
IUPAC Name 4-iodo-3,5-dimethyl-1H-pyrazole
Molecular Weight (g/mol) 222.029
PubChem CID 97115
Formula Weight 222.04
Percent Purity 97%
Chemical Name or Material 4-Iodo-3,5-dimethyl-1H-pyrazole
Show More Show Less
EINECSNumber 622-388-6
RTECSNumber UQ6517000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.