Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-3-methyl-5-phenylisoxazole, 97%, Thermo Scientific™

Catalog No. CC20311EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC20311EA 10 g Amber Glass Bottle
CC20311DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC20311EA Supplier Thermo Scientific Chemicals Supplier No. CC20311EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16114-53-7
Molecular Formula C10H8INO
Molecular Weight (g/mol) 285.084
MDL Number MFCD03086126
InChI Key RLZGZJOMCXKYIW-UHFFFAOYSA-N
Synonym 4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
PubChem CID 2776520
IUPAC Name 4-iodo-3-methyl-5-phenyl-1,2-oxazole
SMILES CC1=NOC(=C1I)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-methyl-5-phenylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.