Learn More
Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 144.0°C to 145.0°C |
| Elemental Analysis | 26.37 to 29.74% (C) 2.21 to 2.50% (H) 6.15 to 6.94% (N) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. as diethylsulfonamide (HPLC) |
| Quantity | 25 g |
| Formula Weight | 214.07 |
| Percent Purity | 95% |
| Chemical Name or Material | Pyridine-3-sulfonyl chloride hydrochloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Causes serious eye damage. Contact with water liberates toxic gas. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.