Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Iodo-4-dimethylaminoazobenzene 97.0+%, TCI America™
SDP

Catalog No. I00575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I00575G Supplier TCI America Supplier No. I00575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3805-67-2
Molecular Formula C14H14IN3
Molecular Weight (g/mol) 351.19
MDL Number MFCD00059492
InChI Key JSCFDMNXBTVUTH-UHFFFAOYSA-N
Synonym 4-Dimethylamino-4′C-iodoazobenzene
PubChem CID 577894
IUPAC Name 4-[2-(4-iodophenyl)diazen-1-yl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(I)C=C1

Specifications

Melting Point 162°C
Quantity 5 g
Formula Weight 351.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Iodo-4-dimethylaminoazobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.