Learn More
Sigma Aldrich 4-Iodo-5-methoxypyridin-3-amine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- N,n-diethylacetoacetamide is a α-dicarbonyl compound that
- Can be used to synthesize potent calcium channel blockers such as 2,6,6-trimethyl-3-diethylaminocarbonyl-4-aryl-5-oxo-1,4,5,6,7,8-hexahydroquinoline derivatives and n,n-diethyl-2,6,6-trimethyl-4-(3-nitrophenyl)-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxamide. it can also undergo michael addition in the presence of bis(2,4-pentanedionato)nickel(ii) catalyst
Specifications
Specifications
| CAS | 1045855-66-0 |
| Molecular Formula | C6H7IN2O |
| Linear Formula | C6H7IN2O |
| MDL Number | MFCD11052847 |
| Quantity | 1 kg |
| Molecular Weight (g/mol) | 250.04 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.