Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-5-methyl-3-phenylisoxazole, 97%, Thermo Scientific™

Catalog No. CC00511EA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00511EA 10 g
CC00511DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00511EA Supplier Thermo Scientific Chemicals Supplier No. CC00511EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31295-66-6
Molecular Formula C10H8INO
Molecular Weight (g/mol) 285.084
MDL Number MFCD02681888
InChI Key DDVPBKUHWSZYIP-UHFFFAOYSA-N
Synonym 4-iodo-5-methyl-3-phenylisoxazole, isoxazole, 4-iodo-5-methyl-3-phenyl, 3-phenyl-4-iodo-5-methylisoxazole
PubChem CID 2776150
IUPAC Name 4-iodo-5-methyl-3-phenyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C2=CC=CC=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 4-Iodo-5-methyl-3-phenylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.