Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Iodoanisole

Catalog No. I7608100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I7608100G 100 g
I760825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I7608100G Supplier Sigma Aldrich Supplier No. I7608100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Iodoanisole

Specifications

CAS 696-62-8
Melting Point 50°C to 53°C (lit.)
Boiling Point 237°C (726 mmHg)
Molecular Formula C7H7IO
Linear Formula IC6H4OCH3
MDL Number MFCD00001056
Quantity 100 g
Synonym 4-Iodophenyl methyl ether; 4-Methoxyiodobenzene
Molecular Weight (g/mol) 234.03
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.