Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodoanisole, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
696-62-8
C7H7IO
234.04
SYSZENVIJHPFNL-UHFFFAOYSA-N
4-iodoanisole,p-iodoanisole,benzene, 1-iodo-4-methoxy,isoform,4-methoxyiodobenzene,p-methoxyiodobenzene,anisole, p-iodo,4-iodomethoxybenzene,p-iodophenyl methyl ether,1-iodo-4-methoxy-benzene
69676
1-iodo-4-methoxybenzene
COC1=CC=C(C=C1)I
696-62-8
C7H7IO
234.04
SYSZENVIJHPFNL-UHFFFAOYSA-N
4-iodoanisole,p-iodoanisole,benzene, 1-iodo-4-methoxy,isoform,4-methoxyiodobenzene,p-methoxyiodobenzene,anisole, p-iodo,4-iodomethoxybenzene,p-iodophenyl methyl ether,1-iodo-4-methoxy-benzene
69676
1-iodo-4-methoxybenzene
COC1=CC=C(C=C1)I
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
Melting Point 46.0°C to 51.0°C
Boiling Point 237.0°C (726.0 mmHg)
Color White to Brown
Flash Point >110°C
Assay Percent Range 98%
Linear Formula IC6H4OCH3
Beilstein 06,208
Formula Weight 234.04
Physical Form Crystalline Powder and Chunks
Percent Purity 98%
Chemical Name or Material 4-Iodoanisole
Show More Show Less

Recommended Storage : product may darken on storage