Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodoanisole 99.0+%, TCI America™
SDP

Catalog No. I0378100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0378100G 100 g
I037825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0378100G Supplier TCI America Supplier No. I0378100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 696-62-8
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001056
InChI Key SYSZENVIJHPFNL-UHFFFAOYSA-N
Synonym 4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene
PubChem CID 69676
IUPAC Name 1-iodo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)I

Specifications

Melting Point 51°C
Boiling Point 240°C
Quantity 100 g
Formula Weight 234.04
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodoanisole
EINECSNumber (3)-3916
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.