Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog No. p-149503
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC412380050 5 g
AC412380250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412380050 Supplier Thermo Scientific Chemicals Supplier No. 412380050
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3058-39-7
Molecular Formula C7H4IN
Molecular Weight (g/mol) 229.02
MDL Number MFCD00051310
InChI Key XOKDXPVXJWTSRM-UHFFFAOYSA-N
Synonym p-iodobenzonitrile, benzonitrile, 4-iodo, p-cyanoiodobenzene, benzonitrile, p-iodo, 1-cyano-4-iodobenzene, 4-cyanoiodobenzene, 4-iodo-benzonitrile, 4-iodobenzenecarbonitrile, 4-iodbenzonitril, 4-iodo benzonitrile
PubChem CID 76467
IUPAC Name 4-iodobenzonitrile
SMILES IC1=CC=C(C=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 125.0°C to 128.0°C
Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 229.02
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Iodobenzonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.