Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodobenzyl Alcohol 99.0+%, TCI America™
SDP

Catalog No. I08105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08105G 5 g
I081025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08105G Supplier TCI America Supplier No. I08105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18282-51-4
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD01732720
InChI Key CNQRHSZYVFYOIE-UHFFFAOYSA-N
Synonym 4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
PubChem CID 29012
IUPAC Name (4-iodophenyl)methanol
SMILES C1=CC(=CC=C1CO)I

Specifications

Melting Point 73°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 234.04
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzyl Alcohol
RTECSNumber DO8318020
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.