Learn More
Medchemexpress LLC 4-iodobenzylamine (4-iodophenyl)methanamine | 39959-59-6 | MFCD00047933 | 99.8% | 233.05 g/mol | C7H8IN | 25 G
SDP
Catalog No. 5000303063
Encompass_Preferred
4-Iodobenzylamine (4-iodophenyl)methanamine is an aryl iodide benzylamine reagent used as a biochemical probe to study serine protease binding (for example, trypsin and urokinase-type plasminogen activator). It is stable in aqueous solution and provided with analytical documentation.
  • Used to probe serine protease binding interactions.
  • Stable in aqueous solutions for assay use.
  • High reported purity for reproducible results.
  • Available in multiple pack sizes for scale flexibility.
  • Supplied with COA and SDS for quality and safety information.
Catalog No. 50-003-03063 Supplier Medchemexpress LLC Supplier No. 25GHYW013964
Add to Cart
Add to Cart
4-Iodobenzylamine (4-iodophenyl)methanamine is an aryl iodide benzylamine reagent used as a biochemical probe to study serine protease binding (for example, trypsin and urokinase-type plasminogen activator). It is stable in aqueous solution and provided with analytical documentation.
  • Used to probe serine protease binding interactions.
  • Stable in aqueous solutions for assay use.
  • High reported purity for reproducible results.
  • Available in multiple pack sizes for scale flexibility.
  • Supplied with COA and SDS for quality and safety information.