Learn More
eMolecules​ 352530-30-4 | 4-Iodobenzylzinc bromide, 0.50 M in THF | Synthonix | MFCD02260148 | 362.310 | C7H6BrIZn | 0.000 | [Br-].[Zn+]Cc1ccc(I)cc1 | 25ml | 627353667
Catalog No. 502695305 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Iodobenzylzinc bromide, 0.50 M in THF | Synthonix | 352530-30-4 | MFCD02260148 | 362.310 | C7H6BrIZn | 0.000 | [Br-].[Zn+]Cc1ccc(I)cc1 | 25ml | 627353667
Catalog No. 50-269-5305 Supplier eMolecules​ Supplier No. 627353667
4-Iodobenzylzinc bromide, 0.50 M in THF | Synthonix | 352530-30-4 | MFCD02260148 | 362.310 | C7H6BrIZn | 0.000 | [Br-].[Zn+]Cc1ccc(I)cc1 | 25ml | 627353667