Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenoxyacetic Acid 97.0+%, TCI America™
SDP

Catalog No. I06445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06445G 5 g
I064425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06445G Supplier TCI America Supplier No. I06445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1878-94-0
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
InChI Key RKORKXFKXYYHAQ-UHFFFAOYSA-N
Synonym 4-iodophenoxyacetic acid, 2-4-iodophenoxy acetic acid, 4-iodophenoxy acetic acid, p-iodophenoxyacetic acid, p-iodophenoxy acetic acid, acetic acid, 4-iodophenoxy, 4-iodophenoxyacetic, 4-iodophenoxyaceticacide, acmc-209eqm, 4-iodo phenoxyacetic acid
PubChem CID 74658
IUPAC Name 2-(4-iodophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)I

Specifications

Melting Point 157°C
Color White
Quantity 5 g
Formula Weight 278.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodophenoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.