Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodotetrahydro-2H-pyran, ≥97%, Thermo Scientific™

Catalog No. CC29911CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC29911CB 250 mg Amber Glass Bottle
CC29911DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC29911CB Supplier Thermo Scientific Chemicals Supplier No. CC29911CB
Add to Cart

Chemical Identifiers

CAS 25637-18-7
Molecular Formula C5H9IO
Molecular Weight (g/mol) 212.03
MDL Number MFCD06797467
InChI Key JTRNQTFTRDPITG-UHFFFAOYSA-N
Synonym 4-iodotetrahydro-2h-pyran, 4-iodo-tetrahydro-2h-pyran, 4-iodotetrahydropyran, 4-iodo-tetrahydropyran, 4-iodo-tetrahydro-pyran, 2h-pyran, tetrahydro-4-iodo, tetrahydro-4-iodo-2h-pyran, 4-iodanyloxane, 4-iodooxacyclohexane, 4-iodopentahydropyran
PubChem CID 2795506
IUPAC Name 4-iodooxane
SMILES IC1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.03
Percent Purity 95%
Chemical Name or Material 4-Iodotetrahydro-2H-pyran
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.