Learn More
Medchemexpress LLC HY-110063 10mg Medchemexpress, 4-IPP CAS:41270-96-6 Purity:>98%
Supplier: Medchemexpress LLC HY11006310MG
Medchemexpress, HY-110063 10mg 4-IPP CAS:41270-96-6 4-IPP (4-Iodo-6-phenylpyrimidine) is a specific suicide substrate and irreversible inhibitor of macrophage migration inhibitory factor (MIF). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.