Learn More
Medchemexpress LLC HY-110063 5mg Medchemexpress, 4-IPP CAS:41270-96-6 Purity:>98%
SDP
Catalog No. 501968739
Encompass_Preferred
Medchemexpress, HY-110063 5mg 4-IPP CAS:41270-96-6 4-IPP (4-Iodo-6-phenylpyrimidine) is a specific suicide substrate and irreversible inhibitor of macrophage migration inhibitory factor (MIF). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8739 Supplier Medchemexpress LLC Supplier No. HY1100635MG
Medchemexpress, HY-110063 5mg 4-IPP CAS:41270-96-6 4-IPP (4-Iodo-6-phenylpyrimidine) is a specific suicide substrate and irreversible inhibitor of macrophage migration inhibitory factor (MIF). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.