Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Isobutylacetophenone, 97%

Catalog No. AC448822500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448822500 250 g
AC448820500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC448822500 Supplier Thermo Scientific Chemicals Supplier No. 448822500
  • $465.00
    $395.65 / Each of 1

    Save $69.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38861-78-8 | C12H16O | 176.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38861-78-8
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.26
InChI Key KEAGRYYGYWZVPC-UHFFFAOYSA-N
Synonym 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
PubChem CID 93214
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanone
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C

Specifications

Density 0.9600g/mL
Boiling Point 107.0°C (2.0 mmHg)
Flash Point 54°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2C6H4COCH3
Quantity 250 g
Specific Gravity 0.96
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 4'-Isobutylacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 254-159-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.