Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Isobutylbenzoic acid | 38861-88-0 | 99.2% | 500 MG
SDP

Catalog No. 5000339847
Encompass_Preferred

4-Isobutylbenzoic acid is an intermediate in the synthesis of compounds. It is intended for research use only and is not for patient administration.

  • Intermediate in chemical synthesis
  • White to off-white solid appearance
  • Molecular formula C11H14O2
  • Molecular weight 178.23
  • Store at -20°C, protected from light

Catalog No. 50-003-39847 Supplier Medchemexpress LLC Supplier No. HY131121500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Isobutylbenzoic acid is an intermediate in the synthesis of compounds. It is intended for research use only and is not for patient administration.

  • Intermediate in chemical synthesis
  • White to off-white solid appearance
  • Molecular formula C11H14O2
  • Molecular weight 178.23
  • Store at -20°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.