Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF

Catalog No. AAH54201AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54201AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54201AE Supplier Thermo Scientific Chemicals Supplier No. H54201AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 863780-45-4 | C10H13BrMg | 237.423 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863780-45-4
Molecular Formula C10H13BrMg
Molecular Weight (g/mol) 237.423
MDL Number MFCD11113503
InChI Key VMAJYUMMJYAXIG-UHFFFAOYSA-M
Synonym p-isobutylphenyl magnesium bromide, qoxyxvturbxklc-uhfffaoysa-m, grignard reagent
PubChem CID 86067760
IUPAC Name magnesium;2-methylpropylbenzene;bromide
SMILES CC(C)CC1=CC=[C-]C=C1.[Mg+2].[Br-]

Specifications

Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 237.42
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 4-Isobutylphenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.