Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole 97+%, Thermo Scientific™

Catalog No. CC15506CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC15506CB 250 mg Amber Glass Bottle
CC15506DA 1 g Amber Glass Bottle
CC15506DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC15506CB Supplier Thermo Scientific Chemicals Supplier No. CC15506CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 799283-97-9
Molecular Formula C11H9N3O
Molecular Weight (g/mol) 199.213
InChI Key IIFVDOYRTLEGHQ-UHFFFAOYSA-N
Synonym 4-isocyanato-5-methyl-1-phenyl-1h-pyrazole, 5-methyl-1-phenylpyrazol-4-isocyanate, 1h-pyrazole,4-isocyanato-5-methyl-1-phenyl, 5-methyl-1-phenyl-1h-pyrazol-4-yl isocyanate, 4-isocyanato-5-methyl-1h-pyrazol-4-yl benzene
PubChem CID 7164551
IUPAC Name 4-isocyanato-5-methyl-1-phenylpyrazole
SMILES CC1=C(C=NN1C2=CC=CC=C2)N=C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-5-methyl-1-phenyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.