Learn More
Cayman Chemical 4-Isocynato TEMPO 10mg
SDP
Catalog No. 502994064
Encompass_Preferred
A spin label for RNA; used to study HIV-1 transactivation response RNA and hammerhead ribosome dynamics by EPR spectroscopy
Catalog No. 50-299-4064 Supplier Cayman Chemical Supplier No. 2307510 MG
A spin label for RNA; used to study HIV-1 transactivation response RNA and hammerhead ribosome dynamics by EPR spectroscopy