Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropoxybenzaldehyde 96.0+%, TCI America™
SDP

Catalog No. I094925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I094925G 25 g
I09495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I094925G Supplier TCI America Supplier No. I094925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18962-05-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00052357
InChI Key WDANSDASCKBVKH-UHFFFAOYSA-N
Synonym 4-isopropoxybenzaldehyde, 4-propan-2-yloxy benzaldehyde, 4-iso-propoxybenzaldehyde, benzaldehyde, 4-1-methylethoxy, 4-propan-2-yl-oxy benzaldehyde, 4-isopropoxy-1-formylbenzene, 4-methylethoxy benzaldehyde, pubchem9651, p-isopropoxybenzaldehyde, 4-isopropoxy-benzaldehyde
PubChem CID 250077
IUPAC Name 4-propan-2-yloxybenzaldehyde
SMILES CC(C)OC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.