Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Isopropoxybenzoic acid | 13205-46-4 | 100.0% | C10H12O3 | 250 MG
SDP

Catalog No. 5000358453
Encompass_Preferred

4-Isopropoxybenzoic acid is an aromatic carboxylic acid organic compound that can be used as a synthetic intermediate in organic synthesis, such as in pesticides. It is a versatile compound with applications in various chemical processes.

  • Aromatic carboxylic acid compound
  • Used as a synthetic intermediate in organic synthesis
  • Molecular weight: 180.20
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-58453 Supplier Medchemexpress LLC Supplier No. 250MGHY75814
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Isopropoxybenzoic acid is an aromatic carboxylic acid organic compound that can be used as a synthetic intermediate in organic synthesis, such as in pesticides. It is a versatile compound with applications in various chemical processes.

  • Aromatic carboxylic acid compound
  • Used as a synthetic intermediate in organic synthesis
  • Molecular weight: 180.20
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.