Learn More
Medchemexpress LLC 4-isopropyl-3-methyl-2-(1-[3-(S)-methyl-piperidin-1-yl]-methanoyl)-2H-isoxalo-5-one | 654059-21-9 | 99.8% | 266.341 g·mol⁻¹ | C14H22N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10205610MG
Encompass_Preferred
BAY 59-9435 is a small-molecule research inhibitor of hormone-sensitive lipase (HSL) used to probe lipolysis and adipocyte lipid metabolism. It is a potent, selective HSL inhibitor (IC50 0.023 μM), supplied as a white to off-white solid and verified for high purity.
  • Potent HSL inhibition (IC50 0.023 μM).
  • High purity suitable for research (HPLC purity ~99.8%).
  • Solid form, white to off-white, easy to handle and weigh.
  • Molecular formula C14H22N2O3; molecular weight 266.341 g·mol⁻¹.
  • Stable under recommended storage conditions (powder: -20°C or 4°C as specified).
  • Available in multiple small-scale quantities for laboratory use.
Catalog No. 50-002-47166