Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylaminodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. P0327500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0327500G 500 g
P032725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0327500G Supplier TCI America Supplier No. P0327500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101-72-4
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD00026348
InChI Key OUBMGJOQLXMSNT-UHFFFAOYSA-N
Synonym ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip
PubChem CID 7573
ChEBI CHEBI:63569
IUPAC Name 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
SMILES CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 80°C
Boiling Point 180°C
Quantity 500 g
UN Number 3077
Formula Weight 226.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Isopropylaminodiphenylamine
EINECSNumber (3)-0136&(3)-0368
RTECSNumber ST2650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.