Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylcyclohexanone, 96%

Catalog No. AAL0886304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0886304 2 g
AAL0886309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0886304 Supplier Thermo Scientific Chemicals Supplier No. L0886304
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5432-85-9 | C9H16O | 140.23 g/mol

4-Isopropylcyclohexanone is used to prepare 4-isopropyl-cyclohexanone oxime. It is also used in the synthesis of dihydroindol-2-ones as agonists and antagonist ligands at the nociceptin receptor. Further, it is used in the preparation of beta-alanine derivatives, which acts as orally available glucagon receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropylcyclohexanone is used to prepare 4-isopropyl-cyclohexanone oxime. It is also used in the synthesis of dihydroindol-2-ones as agonists and antagonist ligands at the nociceptin receptor. Further, it is used in the preparation of beta-alanine derivatives, which acts as orally available glucagon receptor antagonists.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with oxidizing agents. reducing agents and plastics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5432-85-9
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00043479
InChI Key FPKISACHVIIMRA-UHFFFAOYSA-N
Synonym 4-isopropylcyclohexanone, cyclohexanone, 4-1-methylethyl, 4-propan-2-yl cyclohexan-1-one, 4-isopropylcyclohexan-1-one, cyclohexanone, 4-isopropyl, 4-isopropyl-cyclohexanone, 4-iso-propylcyclohexanone, 4-methylethyl cyclohexan-1-one, acmc-20a5fk, 4-isopropyl cyclohexanone
PubChem CID 79488
IUPAC Name 4-propan-2-ylcyclohexan-1-one
SMILES CC(C)C1CCC(=O)CC1

Specifications

Boiling Point 94°C to 96°C (17 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.458
Quantity 2 g
Beilstein 1236952
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 4-Isopropylcyclohexanone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 226-592-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.