Learn More
Medchemexpress LLC 4-isopropylphenyl diphenyl phosphate-d10 | 2512202-12-7 | 98.0% | 378.42 | C21H11D10O4P | 2.5mg
SDP
Catalog No. 5000269407
Encompass_Preferred
4-Isopropylphenyl Diphenyl Phosphate-d10 is the deuterium labeled 4-(1-Methylethyl)phenyl diphenyl phosphate (HY-W588268)
Catalog No. 50-002-69407 Supplier Medchemexpress LLC Supplier No. HYW7134732.5MG
4-Isopropylphenyl Diphenyl Phosphate-d10 is the deuterium labeled 4-(1-Methylethyl)phenyl diphenyl phosphate (HY-W588268)