Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylphenylacetonitrile, 98+%

Catalog No. AAL1071503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1071503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1071503 Supplier Thermo Scientific Chemicals Supplier No. L1071503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4395-87-3 | C11H13N | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4395-87-3
Molecular Formula C11H13N
Molecular Weight (g/mol) 159.23
MDL Number MFCD00040892
InChI Key RIPHZOPMCRSGSI-UHFFFAOYSA-N
Synonym 4-isopropylphenylacetonitrile, 2-4-isopropylphenyl acetonitrile, 4-isopropylphenyl acetonitrile, 2-4-propan-2-yl phenyl acetonitrile, 4-propan-2-yl phenyl acetonitrile, pubchem17248, acmc-20ai20, 2-4-propan-2-ylphenyl acetonitrile, 2-4-methylethyl phenyl ethanenitrile
PubChem CID 138222
IUPAC Name 2-(4-propan-2-ylphenyl)acetonitrile
SMILES CC(C)C1=CC=C(CC#N)C=C1

Specifications

Density 0.96
Boiling Point 150°C (25 mmHg)
Refractive Index 1.512
Quantity 1 g
UN Number UN3276
Beilstein 155941
Formula Weight 159.23
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylphenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.