Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. I06201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0620-1G 1 g
I06205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0620-1G Supplier TCI America Supplier No. I06201G
Add to Cart
Add to Cart

Specifications

CAS 16152-51-5
Melting Point 140°C
Color White-Yellow
Molecular Formula C9H13BO2
Quantity 1 g
MDL Number MFCD01074614
Synonym 4-isopropylphenylboronic acid, 4-isopropylbenzeneboronic acid, 4-isopropylphenyl boronic acid, 4-propan-2-ylphenyl boronic acid, p-isopropylphenylboronic acid
InChI Key IAEUFBDMVKQCLU-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(C)C)(O)O
IUPAC Name (4-propan-2-ylphenyl)boronic acid
Molecular Weight (g/mol) 164.011
PubChem CID 2773480
Formula Weight 164.01
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.